Assay Details
Target Gene Details
Entrez Gene ID: | 320541 |
Gene Name: | solute carrier family 35, member E2 |
Gene Aliases: |
A530082C11Rik, AI957035 |
Location: |
Chr.4:155600592-155623340 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc35e2 | NM_177186.4 | 1 | 103 | NP_796160.1 |
| AK080215.1 | 1 | 119 | ||
| BC058728.1 | 1 | 103 | AAH58728.1 |
More Information
Set Membership: |
|

Genomic Map