Assay Details
Target Gene Details
Entrez Gene ID: | 100502734 |
Gene Name: | predicted gene 16023 |
Gene Aliases: |
OTTMUSG00000029160 |
Location: |
Chr.4:155608297-155624755 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm16023 | NR_040441.1 | 3 | 638 | |
| AK142359.1 | 3 | 639 |
Target Gene Details
Entrez Gene ID: | 320541 |
Gene Name: | solute carrier family 35, member E2 |
Gene Aliases: |
A530082C11Rik, AI957035 |
Location: |
Chr.4:155600592-155623340 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc35e2 | NM_177186.4 | 2 | 637 | NP_796160.1 |
| AK048716.1 | 2 | 506 | BAC33431.1 | |
| AK080215.1 | 2 | 656 | ||
| AK145065.1 | 3 | 560 | BAE26216.1 | |
| AK157354.1 | 2 | 533 | BAE34060.1 | |
| BC058728.1 | 2 | 637 | AAH58728.1 |
More Information
Set Membership: |
|

Genomic Map