Assay Details
Target Gene Details
Entrez Gene ID: | 12164 |
Gene Name: | bone morphogenetic protein 8b |
Gene Aliases: |
Op3 |
Location: |
Chr.4:123104307-123126094 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bmp8b | NM_007559.4 | 1 | 83 | NP_031585.2 |
| AK082895.1 | 1 | 84 | BAC38674.1 | |
| U39545.1 | 1 | 87 | AAB17573.1 |
More Information
Set Membership: |
|

Genomic Map