Assay Details
Target Gene Details
Entrez Gene ID: | 241494 |
Gene Name: | zinc finger protein 385B |
Gene Aliases: |
B830010L13Rik, C130013B13Rik, Zfp533, Znf385b |
Location: |
Chr.2:77410622-77817706 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp385b | NM_178723.6 | 1 | 131 | NP_848838.2 |
| AK033075.1 | BAC28144.1 | |||
| AK046798.1 | 1 | 132 | BAC32874.1 | |
| AK079322.1 | ||||
| AK149352.1 | 1 | 114 | BAE28828.1 |

Genomic Map