Assay Details
Target Gene Details
Entrez Gene ID: | 99542 |
Gene Name: | expressed sequence AI115009 |
Gene Aliases: |
- |
Location: |
Chr.3:152666913-152681432 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AI115009 | NR_040386.1 | |||
| AK043786.1 |
Target Gene Details
Entrez Gene ID: | 229949 |
Gene Name: | adenylate kinase 5 |
Gene Aliases: |
- |
Location: |
Chr.3:152462815-152668376 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ak5 | NM_001081277.1 | 1 | 177 | NP_001074746.1 |
| AK053807.1 | 1 | 177 |
More Information
Set Membership: |
|

Genomic Map