Assay Details
Target Gene Details
Entrez Gene ID: | 13639 |
Gene Name: | ephrin A4 |
Gene Aliases: |
EFL-4, Epl4, LERK-4 |
Location: |
Chr.3:89333393-89338028 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Efna4 | NM_007910.2 | 4 | 732 | NP_031936.2 |
| AK012195.1 | 4 | 736 | BAB28092.1 | |
| AK142604.1 | 4 | 733 | BAE25127.1 | |
| U90663.1 | 4 | 732 | AAB50238.1 |
More Information
Set Membership: |
|

Genomic Map