Assay Details
Target Gene Details
Entrez Gene ID: | 75812 |
Gene Name: | taspase, threonine aspartase 1 |
Gene Aliases: |
4930485D02Rik, AW986064 |
Location: |
Chr.2:139833479-140067666 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tasp1 | NM_001159640.2 | 14 | 1455 | NP_001153112.1 |
| NM_001159641.2 | 13 | 1344 | NP_001153113.1 | |
| NM_175225.5 | 14 | 1722 | NP_780434.1 | |
| NR_110350.1 | 13 | 1629 | ||
| NR_110351.1 | 14 | 1442 | ||
| NR_110352.1 | 13 | 1373 | ||
| AK015624.1 | 14 | 1442 | ||
| AK036512.1 | 14 | 1678 | BAC29457.1 | |
| AK132709.1 | 13 | 1359 | ||
| AK146320.1 | 13 | 1327 | BAE27075.1 | |
| AK160983.1 | 13 | 1590 | ||
| BC024597.1 | 14 | 1442 | AAH24597.1 |
More Information
Set Membership: |
|

Genomic Map