Assay Details
Target Gene Details
Entrez Gene ID: | 20333 |
Gene Name: | SEC22 vesicle trafficking protein homolog B (S. cerevisiae) |
Gene Aliases: |
4930564D15Rik, AA517334, AI480645, C81333, ERS-24, Sec22l1 |
Location: |
Chr.3:97901227-97922318 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sec22b | NM_011342.4 | 5 | 1616 | NP_035472.1 |
| AK037918.1 | 5 | 1617 | BAC29901.1 |

Genomic Map