Assay Details
Target Gene Details
Entrez Gene ID: | 211550 |
Gene Name: | TRAF-interacting protein with forkhead-associated domain |
Gene Aliases: |
T2bp |
Location: |
Chr.3:127788857-127798394 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tifa | NM_145133.3 | 1 | 208 | NP_660115.1 |
| AB062111.1 | BAB86903.1 | |||
| AK044221.1 | 1 | 209 | BAC31825.1 | |
| AK149864.1 | 1 | 176 | BAE29133.1 | |
| AK151651.1 | 1 | 159 | BAE30580.1 | |
| AK166239.1 | 1 | 250 | BAE38653.1 | |
| BC065775.1 | 1 | 202 | AAH65775.1 | |
| BY168652.1 | 1 | 208 |

Genomic Map