Assay Details
Target Gene Details
Entrez Gene ID: | 109676 |
Gene Name: | ankyrin 2, brain |
Gene Aliases: |
AI835472, AW491075, Ank-2 |
Location: |
Chr.3:126921607-127499870 on Build GRCm38 |
Assay Gene Location: | Within Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ank2 | NM_001034168.1 | 10 | 3889 | NP_001029340.1 |
| NM_178655.3 | 25 | 4192 | NP_848770.2 | |
| AK032060.1 | 25 | 4192 | BAC27676.1 | |
| AK050264.1 | 1 | 788 | ||
| AK083826.1 | 10 | 2448 | BAE43385.1 | |
| BC043123.1 | 10 | 3889 | AAH43123.1 | |
| BC059251.1 | 25 | 4143 | AAH59251.1 |
More Information
Set Membership: |
|

Genomic Map