Assay Details
Target Gene Details
Entrez Gene ID: | 66868 |
Gene Name: | major facilitator superfamily domain containing 1 |
Gene Aliases: |
1200003O06Rik |
Location: |
Chr.3:67582768-67604231 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mfsd1 | NM_025813.3 | 16 | 2723 | NP_080089.1 |
| AK004586.1 | 16 | 2724 | BAB23391.1 | |
| AK032688.1 | 16 | 2691 | ||
| AK135235.1 | 16 | 2724 | BAE22467.1 | |
| AK171855.1 | 1 | 836 | BAE42699.1 | |
| BC024891.1 | 16 | 2705 | AAH24891.1 |

Genomic Map