Assay Details
Target Gene Details
Entrez Gene ID: | 226548 |
Gene Name: | anterior pharynx defective 1a homolog (C. elegans) |
Gene Aliases: |
6530402N02Rik, APH-1a |
Location: |
Chr.3:95893907-95898592 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aph1a | NM_146104.3 | 6 | 2586 | NP_666216.1 |
| NM_146134.2 | 7 | 2253 | NP_666246.1 | |
| AK148400.1 | 6 | 2539 | BAE28531.1 | |
| AK190617.1 | 1 | 339 | ||
| AK206190.1 | 1 | 70 | ||
| BC012406.1 | 6 | 2511 | AAH12406.1 |
Target Gene Details
Entrez Gene ID: | 23831 |
Gene Name: | carbonic anhydrase 14 |
Gene Aliases: |
AW536446, Ca14 |
Location: |
Chr.3:95897768-95905118 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Car14 | NM_011797.2 | 11 | 1462 | NP_035927.1 |
| AB005450.1 | 11 | 1462 | BAA78709.1 | |
| AK012316.1 | 8 | 839 | ||
| AK157764.1 | 11 | 1463 | ||
| BC046995.1 | 11 | 1444 | AAH46995.1 |
More Information
Set Membership: |
|

Genomic Map