Assay Details
Target Gene Details
Entrez Gene ID: | 246703 |
Gene Name: | apolipoprotein A-I binding protein |
Gene Aliases: |
AA087124, AI-BP, AIBP, Apoa1ip, ESTM37 |
Location: |
Chr.3:88056523-88058541 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Apoa1bp | NM_144897.3 | 6 | 745 | NP_659146.1 |
| AJ344092.1 | 6 | 745 | CAC86966.1 | |
| AK131615.1 | 6 | 956 | ||
| AK159846.1 | 6 | 747 | BAE35424.1 | |
| AY566271.1 | 6 | 745 | AAT70236.1 | |
| BC058362.1 | 6 | 743 | AAH58362.1 |
More Information
Set Membership: |
|

Genomic Map