Assay Details
Target Gene Details
Entrez Gene ID: | 80890 |
Gene Name: | tripartite motif-containing 2 |
Gene Aliases: |
mKIAA0517, narf |
Location: |
Chr.3:84160439-84305832 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trim2 | NM_001271725.1 | 12 | 3410 | NP_001258654.1 |
| NM_001271726.1 | 12 | 3446 | NP_001258655.1 | |
| NM_001271727.1 | 12 | 3715 | NP_001258656.1 | |
| NM_001271728.1 | 12 | 3513 | NP_001258657.1 | |
| NM_030706.3 | 12 | 3509 | NP_109631.2 | |
| AB043550.1 | 12 | 3493 | BAB17634.1 | |
| AF220017.1 | 12 | 3358 | AAG53471.1 | |
| AK042752.1 | 4 | 1502 | BAC31352.1 | |
| AK045410.1 | 12 | 3506 | BAC32350.1 | |
| AK079415.1 | 12 | 3660 | BAC37640.1 | |
| AK141419.1 | 12 | 3356 | BAE24679.1 | |
| AK147275.1 | 12 | 3623 | BAE27812.1 | |
| AK147395.1 | 12 | 3504 | BAE27883.1 | |
| AK164350.1 | 11 | 3212 | BAE37753.1 | |
| AK220343.1 | 11 | 4952 | BAD90242.1 | |
| BC058961.1 | 12 | 3513 | AAH58961.1 |
More Information
Set Membership: |
|

Genomic Map