Assay Details
Target Gene Details
Entrez Gene ID: | 212647 |
Gene Name: | aldehyde dehydrogenase 4 family, member A1 |
Gene Aliases: |
A930035F14Rik, Ahd-1, Ahd1, Aldh4, Aldh5a1, E330022C09, P5cd, P5cdh, P5cdhl, P5cdhs, Ssdh1 |
Location: |
Chr.4:139622871-139649691 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aldh4a1 | NM_175438.4 | 15 | 2326 | NP_780647.3 |
| AK044712.1 | 15 | 2122 | BAC32045.1 | |
| AK087796.1 | 2 | 526 | BAC40006.1 | |
| BC024133.1 | 14 | 2044 | AAH24133.1 | |
| BC026589.1 | 10 | 1538 | AAH26589.1 | |
| BC039281.1 | 15 | 2110 | AAH39281.2 | |
| BC056226.1 | 15 | 2210 | AAH56226.1 | |
| BC060650.1 | AAH60650.1 |
More Information
Set Membership: |
|

Genomic Map