Assay Details
Target Gene Details
Entrez Gene ID: | 30933 |
Gene Name: | torsin family 2, member A |
Gene Aliases: |
Prosalusin |
Location: |
Chr.2:32757231-32762244 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tor2a | NM_152800.3 | 2 | 302 | NP_690013.1 |
| NR_045485.1 | ||||
| AK019246.1 | BAC25585.1 | |||
| AK162719.1 | 2 | 246 | BAE37036.1 | |
| AK170979.1 | 1 | 1481 | BAE42156.1 | |
| AK171247.1 | 2 | 254 | BAE42340.1 | |
| AK185253.1 | 2 | 150 | ||
| AK189495.1 | ||||
| BC003466.1 | 2 | 287 | AAH03466.1 | |
| BC024469.1 | 2 | 265 | AAH24469.1 | |
| BY052117.1 | ||||
| CK031473.1 |
Target Gene Details
Entrez Gene ID: | 338348 |
Gene Name: | tetratricopeptide repeat domain 16 |
Gene Aliases: |
1200002K10Rik |
Location: |
Chr.2:32757026-32775663 on Build GRCm38 |
Assay Gene Location: | Within Intron 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttc16 | NM_001290563.1 | NP_001277492.1 | ||
| AK160435.1 | BAE35787.1 |
More Information
Set Membership: |
|

Genomic Map