Assay Details
Target Gene Details
Entrez Gene ID: | 242083 |
Gene Name: | protein phosphatase 1 (formerly 2C)-like |
Gene Aliases: |
3222401G13, 5930404J21Rik, AI481720, AW045850, PP2C, PP2Cepsilon, Pp2ce, Ppp2ce, mKIAA4175 |
Location: |
Chr.3:69316918-69560800 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppm1l | NM_178726.3 | 1 | 872 | NP_848841.2 |
| AK032529.1 | 1 | 930 | BAC27913.1 | |
| AK035912.1 | 1 | 875 | BAC29241.1 | |
| AK045724.1 | 1 | 913 | BAC32472.1 | |
| AK131646.1 | 1 | 859 | BAE20738.1 | |
| AK142789.1 | 1 | 890 | ||
| AK147389.1 | 1 | 875 | ||
| AK147876.1 | 1 | 908 | BAE28196.1 | |
| AK165160.1 | 1 | 905 | ||
| AK220523.1 | 1 | 877 | BAD90308.1 | |
| AY184801.1 | 1 | 460 | AAO43055.1 | |
| BC096031.1 | 1 | 872 | AAH96031.1 |
More Information
Set Membership: |
|

Genomic Map