Assay Details
Target Gene Details
Entrez Gene ID: | 242083 |
Gene Name: | protein phosphatase 1 (formerly 2C)-like |
Gene Aliases: |
3222401G13, 5930404J21Rik, AI481720, AW045850, PP2C, PP2Cepsilon, Pp2ce, Ppp2ce, mKIAA4175 |
Location: |
Chr.3:69316918-69560800 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppm1l | NM_178726.3 | 4 | 2671 | NP_848841.2 |
| AK028275.1 | 4 | 1713 | BAC25853.1 | |
| AK032529.1 | 4 | 2729 | BAC27913.1 | |
| AK035912.1 | 4 | 2674 | BAC29241.1 | |
| AK045724.1 | 4 | 2712 | BAC32472.1 | |
| AK131646.1 | 4 | 2658 | BAE20738.1 | |
| AK142789.1 | 4 | 2690 | ||
| AK147389.1 | 4 | 2672 | ||
| AK147876.1 | 4 | 2708 | BAE28196.1 | |
| AK165160.1 | 4 | 2704 | ||
| AK220523.1 | 4 | 2676 | BAD90308.1 | |
| BC096031.1 | 4 | 2671 | AAH96031.1 |
More Information
Set Membership: |
|

Genomic Map