Assay Details
Target Gene Details
Entrez Gene ID: | 381373 |
Gene Name: | trans-acting transcription factor 9 |
Gene Aliases: |
- |
Location: |
Chr.2:73271926-73275771 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sp9 | NM_001005343.2 | 2 | 1930 | NP_001005343.1 |
| AK135123.1 | 2 | 1925 | ||
| AY591908.1 | 2 | 1940 | AAU04517.1 | |
| BC139398.1 | 2 | 1854 | ||
| BC139407.1 | 2 | 1854 |
More Information
Set Membership: |
|

Genomic Map