Assay Details
Target Gene Details
Entrez Gene ID: | 56224 |
Gene Name: | tetraspanin 5 |
Gene Aliases: |
2810455A09Rik, 4930505M03Rik, NET-4, Tm4sf9 |
Location: |
Chr.3:138742208-138904433 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tspan5 | NM_019571.5 | 8 | 1458 | NP_062517.1 |
| NR_121195.1 | 8 | 1394 | ||
| AF121344.1 | 8 | 1473 | AAF28869.1 | |
| AK013350.1 | 8 | 1423 | BAB28804.1 | |
| AK033543.1 | 8 | 1157 | ||
| AK182422.1 | ||||
| AK190582.1 | 1 | 49 | ||
| AK195572.1 | 1 | 49 | ||
| BC046784.1 | 8 | 1297 | ||
| BC058695.1 | 8 | 1429 | AAH58695.1 |

Genomic Map