Assay Details
Target Gene Details
Entrez Gene ID: | 67513 |
Gene Name: | RIKEN cDNA 2610002J02 gene |
Gene Aliases: |
Faap20 |
Location: |
Chr.4:155246759-155256687 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 2610002J02Rik | NM_001190445.1 | 4 | 703 | NP_001177374.1 |
| NR_033802.1 | 3 | 1146 | ||
| AK003268.1 | 4 | 697 | ||
| AK011291.1 | 4 | 703 | ||
| AK144436.1 | 3 | 1117 | BAE25889.1 | |
| AK219952.1 | ||||
| BC117034.1 | 4 | 689 |

Genomic Map