Assay Details
Target Gene Details
Entrez Gene ID: | 72007 |
Gene Name: | fibronectin type III domain containing 3B |
Gene Aliases: |
1600019O04Rik, AW550168, Fad104, mKIAA4164 |
Location: |
Chr.3:27416161-27711662 on Build GRCm38 |
Assay Gene Location: | Within Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fndc3b | NM_173182.2 | 26 | 5157 | NP_775274.2 |
| AB098596.1 | 26 | 5095 | BAC53726.1 | |
| AK005500.1 | 1 | 1359 | ||
| AK050037.1 | ||||
| AK147346.1 | 27 | 5167 | ||
| AK220297.1 | 26 | 5157 | BAD90222.1 | |
| BC067389.1 | 26 | 5163 | AAH67389.1 | |
| DQ192037.1 | 25 | 5063 | ABA82150.1 |
More Information
Set Membership: |
|

Genomic Map