Assay Details
Target Gene Details
Entrez Gene ID: | 629756 |
Gene Name: | WAP four-disulfide core domain 10 |
Gene Aliases: |
Gm120, OTTMUSG00000016289 |
Location: |
Chr.2:164656046-164657368 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Wfdc10 | NM_001039501.2 | 2 | 309 | NP_001034590.1 |
| AK131754.1 | 2 | 310 | BAE20798.1 | |
| BC132270.1 | 2 | 300 | ||
| BC132272.1 | 2 | 300 | ||
| DQ437333.1 | 2 | 415 |
More Information
Set Membership: |
|

Genomic Map