Assay Details
Target Gene Details
Entrez Gene ID: | 100041480 |
Gene Name: | myelin basic protein expression factor 2, repressor pseudogene |
Gene Aliases: |
ENSMUSG00000049230 |
Location: |
Chr.3:10088277-10092562 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm9833 | NR_045710.1 | 1 | 229 | |
| BC060946.1 | 1 | 229 | AAH60946.1 |
More Information
Set Membership: |
|

Genomic Map