Assay Details
Target Gene Details
Entrez Gene ID: | 70599 |
Gene Name: | sperm specific antigen 2 |
Gene Aliases: |
5730488C15Rik, CS-1, CS1, KRAP, SPAG13, mKIAA1927 |
Location: |
Chr.2:79635373-79672966 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ssfa2 | NM_080558.4 | 1 | 158 | NP_542125.3 |
| AB093303.1 | BAC41485.1 | |||
| AB120565.1 | BAC87833.1 | |||
| AK169543.1 | 1 | 159 | BAE41220.1 | |
| BC115799.1 | ||||
| BC122519.1 | ||||
| BC141883.1 | ||||
| CF539296.1 | ||||
| CJ129385.1 | 1 | 207 |

Genomic Map