Assay Details
Target Gene Details
Entrez Gene ID: | 70599 |
Gene Name: | sperm specific antigen 2 |
Gene Aliases: |
5730488C15Rik, CS-1, CS1, KRAP, SPAG13, mKIAA1927 |
Location: |
Chr.2:79635373-79672966 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ssfa2 | NM_080558.4 | 16 | 3386 | NP_542125.3 |
| AB093303.1 | 16 | 3261 | BAC41485.1 | |
| AB120565.1 | 16 | 3271 | BAC87833.1 | |
| AK037213.1 | 14 | 3096 | BAC29756.1 | |
| AK169543.1 | 17 | 3564 | BAE41220.1 | |
| BC008560.1 | 9 | 2359 | AAH08560.1 | |
| BC018526.1 | 6 | 590 | ||
| BC079535.1 | 10 | 2765 | AAH79535.1 | |
| BC115799.1 | 3 | 172 | ||
| BC122519.1 | 16 | 3272 | ||
| BC141883.1 | 16 | 3271 |

Genomic Map