Assay Details
Target Gene Details
Entrez Gene ID: | 213262 |
Gene Name: | follistatin-like 5 |
Gene Aliases: |
9130207J01Rik |
Location: |
Chr.3:76074275-76710010 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fstl5 | NM_178673.4 | 1 | 103 | NP_848788.2 |
| AF374460.1 | 1 | 77 | AAQ02778.1 | |
| AK033652.1 | 1 | 94 | BAC28408.1 | |
| AK081152.1 | 1 | 94 | BAC38146.1 | |
| AK138388.1 | 1 | 395 | BAE23641.1 | |
| AK140611.1 | 1 | 106 | BAE24428.1 | |
| BB646895.1 | 1 | 103 | ||
| BC075640.1 | 1 | 66 | AAH75640.1 |
More Information
Set Membership: |
|

Genomic Map