Assay Details
Target Gene Details
Entrez Gene ID: | 140917 |
Gene Name: | DNA cross-link repair 1B, PSO2 homolog (S. cerevisiae) |
Gene Aliases: |
AI452214, Apollo, SNMIB, mSNM1B |
Location: |
Chr.3:103800605-103809387 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dclre1b | NM_001025312.1 | 1 | 326 | NP_001020483.1 |
| NM_133865.2 | 1 | 326 | NP_598626.2 | |
| AK046571.1 | 1 | 329 | BAC32791.1 | |
| AK049115.1 | 1 | 321 | BAC33550.1 | |
| AK051104.1 | 1 | 281 | ||
| AK084347.1 | 1 | 320 | BAC39165.1 | |
| AK134324.1 | 1 | 86 | BAE22098.1 | |
| AK138568.1 | 1 | 384 | BAE23700.1 | |
| AK154124.1 | 1 | 229 | BAE32389.1 | |
| AK163827.1 | 1 | 265 | ||
| BC067017.1 | 1 | 235 | AAH67017.1 | |
| BQ559794.1 | 1 | 326 | ||
| CA548899.1 | 1 | 326 |
More Information
Set Membership: |
|

Genomic Map