Assay Details
Target Gene Details
Entrez Gene ID: | 140917 |
Gene Name: | DNA cross-link repair 1B, PSO2 homolog (S. cerevisiae) |
Gene Aliases: |
AI452214, Apollo, SNMIB, mSNM1B |
Location: |
Chr.3:103800605-103809387 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dclre1b | NM_001025312.1 | 3 | 1244 | NP_001020483.1 |
| NM_133865.2 | 4 | 1410 | NP_598626.2 | |
| AK049115.1 | 4 | 1405 | BAC33550.1 | |
| AK051104.1 | 4 | 1365 | ||
| AK084347.1 | 3 | 1239 | BAC39165.1 | |
| AK134324.1 | 4 | 1170 | BAE22098.1 | |
| AK137495.1 | 4 | 967 | BAE23379.1 | |
| AK138568.1 | 4 | 1468 | BAE23700.1 | |
| AK154124.1 | 4 | 1312 | BAE32389.1 | |
| BC011094.1 | 3 | 869 | AAH11094.1 | |
| BC067017.1 | 3 | 1153 | AAH67017.1 | |
| BC125277.1 | 3 | 865 | ||
| CA317555.1 | 1 | 129 |
Target Gene Details
Entrez Gene ID: | 100503157 |
Gene Name: | predicted gene 15471 |
Gene Aliases: |
OTTMUSG00000022448 |
Location: |
Chr.3:103803129-103808440 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm15471 | NR_040412.1 | |||
| AK052792.1 | ||||
| BX516086.1 |
More Information
Set Membership: |
|

Genomic Map