Assay Details
Target Gene Details
Entrez Gene ID: | 22329 |
Gene Name: | vascular cell adhesion molecule 1 |
Gene Aliases: |
CD106, Vcam-1 |
Location: |
Chr.3:116110020-116129688 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Vcam1 | NM_011693.3 | 4 | 1077 | NP_035823.3 |
| AK016465.1 | 4 | 846 | ||
| AK030195.1 | 4 | 847 | BAC26834.1 | |
| AK143401.1 | 4 | 847 | BAE25364.1 | |
| AK162954.1 | 4 | 847 | BAE37132.1 | |
| BC011159.1 | 4 | 819 | AAH11159.2 | |
| BC029823.1 | 4 | 1077 | AAH29823.1 | |
| CT010373.1 | 4 | 756 | CAJ18580.1 | |
| L08431.1 | 4 | 816 | AAA40546.1 | |
| M84487.1 | 4 | 813 | AAA40545.1 | |
| U12884.1 | 4 | 799 | AAA64832.1 | |
| X67783.1 | 4 | 771 | CAA47989.1 |

Genomic Map