Assay Details
Target Gene Details
Entrez Gene ID: | 108058 |
Gene Name: | calcium/calmodulin-dependent protein kinase II, delta |
Gene Aliases: |
2810011D23Rik, 8030469K03Rik, CaMK II, [d]-CaMKII |
Location: |
Chr.3:126596358-126846326 on Build GRCm38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Camk2d | NM_001025438.2 | 19 | 3532 | NP_001020609.1 |
| NM_001025439.2 | 20 | 3545 | NP_001020610.1 | |
| NM_001293663.1 | 20 | 3545 | NP_001280592.1 | |
| NM_001293664.1 | 19 | 3485 | NP_001280593.1 | |
| NM_001293665.1 | 20 | 3525 | NP_001280594.1 | |
| NM_023813.4 | 18 | 3443 | NP_076302.1 | |
| AK032130.1 | 1 | 909 | ||
| AK132185.1 | 5 | 1962 | BAE21020.1 | |
| AK164291.1 | 1 | 881 | ||
| AK220516.1 | 20 | 3545 | BAD90304.1 | |
| BC052894.1 | 19 | 3143 | AAH52894.1 |
More Information
Set Membership: |
|

Genomic Map