Assay Details
Target Gene Details
Entrez Gene ID: | 12349 |
Gene Name: | carbonic anhydrase 2 |
Gene Aliases: |
AI131712, CAII, Ca2, Car-2, Ltw-5, Lvtw-5 |
Location: |
Chr.3:14886278-14900770 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Car2 | NM_009801.4 | 1 | 183 | NP_033931.4 |
BU946040.1 | 1 | 185 | ||
U37705.1 | 1 | 157 |
