Assay Details
Target Gene Details
Entrez Gene ID: | 14616 |
Gene Name: | gap junction protein, alpha 8 |
Gene Aliases: |
Aey5, Cnx50, Cx50, Lop10 |
Location: |
Chr.3:96913566-96926051 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gja8 | NM_008123.3 | 2 | 628 | NP_032149.1 |
| BB665296.1 | 2 | 507 | ||
| BC120513.1 | 2 | 549 | ||
| BC125569.1 | 2 | 549 | ||
| GU363513.1 | 2 | 507 | ||
| GU363514.1 | 2 | 597 | ||
| GU363515.1 | 1 | 814 |
More Information
Set Membership: |
|

Genomic Map