Assay Details
Target Gene Details
Entrez Gene ID: | 320302 |
Gene Name: | glycosyltransferase 28 domain containing 2 |
Gene Aliases: |
4732486J07Rik, Glt28d1l |
Location: |
Chr.3:85869846-85887518 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Glt28d2 | NM_177130.3 | 1 | 303 | NP_796104.2 |
| AK029059.1 | 1 | 179 | BAC26271.1 | |
| AK030588.1 | 1 | 192 | BAC27032.1 | |
| BC119772.1 | ||||
| BC121823.1 |
More Information
Set Membership: |
|

Genomic Map