Assay Details
Target Gene Details
| Entrez Gene ID: | 329650 | 
| Gene Name: | mediator complex subunit 12-like | 
| Gene Aliases: | A130035F20, A630079M02, AU044581, TRALPUSH | 
| Location: | Chr.3:59005595-59318416 on Build GRCm38 | 
| Assay Gene Location: | Within Intron 15 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| Med12l | NM_177855.3 | NP_808523.2 | ||
| AK047743.1 | BAC33144.1 | |||
| AK129475.1 | 16 | 2754 | BAC98285.1 | |
| BP757618.1 | 
Target Gene Details
| Entrez Gene ID: | 140795 | 
| Gene Name: | purinergic receptor P2Y, G-protein coupled, 14 | 
| Gene Aliases: | A330108O13Rik, Gpr105, P2Y14 | 
| Location: | Chr.3:59114623-59153492 on Build GRCm38 | 
| Assay Gene Location: | Within Exon 5 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| P2ry14 | NM_001008497.2 | 3 | 860 | NP_001008497.1 | 
| NM_001287119.1 | 3 | 665 | NP_001274048.1 | |
| NM_001287120.1 | 2 | 790 | NP_001274049.1 | |
| NM_001287121.1 | 3 | 916 | NP_001274050.1 | |
| NM_001287122.1 | 2 | 993 | NP_001274051.1 | |
| NM_001287123.1 | 2 | 1042 | NP_001274052.1 | |
| NM_001287124.1 | 2 | 756 | NP_001274053.1 | |
| NM_133200.4 | 2 | 986 | NP_573463.1 | |
| AF177211.1 | 2 | 847 | AAG09275.1 | |
| AK039791.1 | 2 | 1044 | BAC30456.1 | |
| AK140301.1 | 2 | 818 | BAE24324.1 | |
| AK155035.1 | 3 | 668 | ||
| AK164982.1 | 2 | 805 | BAE37990.1 | |
| BC028995.1 | 2 | 795 | AAH28995.1 | |
| BC058558.1 | 3 | 722 | AAH58558.1 | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Exonic  Intronic  Non-exonic | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top