Assay Details
Target Gene Details
Entrez Gene ID: | 229700 |
Gene Name: | RNA binding motif protein 15 |
Gene Aliases: |
C230088J01Rik, mKIAA1438 |
Location: |
Chr.3:107326110-107333289 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rbm15 | NM_001045807.1 | 1 | 2558 | NP_001039272.1 |
| AK077048.1 | 1 | 2943 | ||
| AK166271.1 | 1 | 2321 | BAE38672.1 | |
| AK168242.1 | 1 | 2319 | BAE40192.1 | |
| AK168936.1 | 1 | 2443 | ||
| AK220162.1 | 1 | 2370 | BAD90348.1 | |
| BC028452.1 | 1 | 116 | ||
| BC051409.1 | 1 | 2312 | AAH51409.1 | |
| BC057038.1 | 1 | 2322 | AAH57038.1 | |
| BC080828.1 | 1 | 2303 | AAH80828.1 | |
| BC120590.1 | 1 | 2558 | ||
| BC137741.1 | 1 | 2558 |
More Information
Set Membership: |
|

Genomic Map