Assay Details
Target Gene Details
Entrez Gene ID: | 12017 |
Gene Name: | BCL2-associated athanogene 1 |
Gene Aliases: |
BAG-1, Rap46 |
Location: |
Chr.4:40936398-40948294 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Bag1 | NM_001171739.1 | 1 | 293 | NP_001165210.1 |
NM_009736.3 | 1 | 293 | NP_033866.4 | |
AF019889.1 | 1 | 79 | ||
AF022223.1 | 1 | 234 | AAC34259.1 | |
AK009149.1 | 1 | 206 | BAB26106.1 | |
AK154469.1 | 1 | 243 | ||
AK171094.1 | 1 | 243 | ||
AK195259.1 | 1 | 87 | ||
BC003722.1 | 1 | 255 | ||
BC069918.1 | 1 | 230 | ||
BC093509.1 | 1 | 204 | ||
CA321856.1 | 1 | 280 | ||
CX225744.1 | 1 | 293 |
