Assay Details
Target Gene Details
Entrez Gene ID: | 67333 |
Gene Name: | serine/threonine kinase 35 |
Gene Aliases: |
1700054C12Rik, CLIK1, Stk35l1 |
Location: |
Chr.2:129800517-129832287 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Stk35 | NM_001038635.2 | NP_001033724.2 | ||
| NM_183262.3 | 3 | 1056 | NP_899085.3 | |
| AK006778.1 | 3 | 821 | BAC25152.1 | |
| AK165988.1 | BAE38504.1 | |||
| BC047277.1 | 2 | 576 | AAH47277.1 | |
| BC138904.1 | 3 | 963 | ||
| BC138905.1 | 3 | 960 |
More Information
Set Membership: |
|

Genomic Map