Assay Details
Target Gene Details
Entrez Gene ID: | 100042889 |
Gene Name: | RIKEN cDNA C920021L13 gene |
Gene Aliases: |
- |
Location: |
Chr.3:95871522-95889093 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C920021L13Rik | NR_040446.1 | |||
| AK044447.1 | ||||
| AK160363.1 |
Target Gene Details
Entrez Gene ID: | 229599 |
Gene Name: | circadian associated repressor of transcription |
Gene Aliases: |
Chrono, Gm129 |
Location: |
Chr.3:95878500-95882421 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ciart | NM_001033302.2 | 6 | 1293 | NP_001028474.1 |
| AK156084.1 | 5 | 1835 | BAE33580.1 | |
| AK164001.1 | 6 | 1293 | BAE37582.1 | |
| BC132471.1 | ||||
| BC138510.1 |
More Information
Set Membership: |
|

Genomic Map