Assay Details
Target Gene Details
Entrez Gene ID: | 242235 |
Gene Name: | leucine-rich repeat, immunoglobulin-like and transmembrane domains 3 |
Gene Aliases: |
Gm421 |
Location: |
Chr.3:129786929-129808884 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lrit3 |
Target Gene Details
Entrez Gene ID: | 20132 |
Gene Name: | retinal pigment epithelium derived rhodopsin homolog |
Gene Aliases: |
- |
Location: |
Chr.3:129808575-129822505 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rrh | NM_009102.3 | 7 | 1165 | NP_033128.1 |
| AF012271.1 | AAC53344.1 | |||
| AK021379.1 | 5 | 876 | BAB32391.1 | |
| BC046288.1 | 8 | 1291 | AAH46288.1 |
More Information
Set Membership: |
|

Genomic Map