Assay Details
Target Gene Details
Entrez Gene ID: | 227525 |
Gene Name: | DNA cross-link repair 1C, PSO2 homolog (S. cerevisiae) |
Gene Aliases: |
9930121L06Rik, AI661365, Art, Snm1l |
Location: |
Chr.2:3424062-3474330 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dclre1c |
Target Gene Details
Entrez Gene ID: | 64707 |
Gene Name: | suppressor of variegation 3-9 homolog 2 (Drosophila) |
Gene Aliases: |
4930507K23Rik, AA536750, D030054H19Rik, D2Ertd544e, KMT1B |
Location: |
Chr.2:3455815-3474986 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Suv39h2 | NM_022724.4 | 3 | 866 | NP_073561.2 |
| NR_027509.1 | 2 | 720 | ||
| AF149205.1 | 3 | 844 | AAG09134.1 | |
| AK045206.1 | 2 | 704 | ||
| AK083457.1 | 3 | 844 | BAC38921.1 | |
| AK165278.1 | 2 | 519 | BAE38116.1 | |
| BC032960.1 | AAH32960.1 |
More Information
Set Membership: |
|

Genomic Map