Assay Details
Target Gene Details
Entrez Gene ID: | 242384 |
Gene Name: | leucine rich repeat and Ig domain containing 2 |
Gene Aliases: |
B230217C06Rik, Lern3, Lrrn6c |
Location: |
Chr.4:35706647-36952254 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lingo2 | NM_001165999.1 | 8 | 2145 | NP_001159471.1 |
| NM_001166000.1 | 9 | 1604 | NP_001159472.1 | |
| NM_001166001.1 | 2 | 1065 | NP_001159473.1 | |
| NM_175516.4 | 3 | 1172 | NP_780725.2 | |
| AK035778.1 | 3 | 1114 | BAC29183.1 | |
| AK045639.1 | 3 | 1173 | BAC32442.1 | |
| AK141568.1 | 8 | 1628 | BAE24739.1 | |
| BC090619.1 | 9 | 1532 | ||
| BC137866.1 | 2 | 1006 | ||
| BC144985.1 | 2 | 1006 | ||
| BC145692.1 | 2 | 1006 |
More Information
Set Membership: |
|

Genomic Map