Assay Details
Target Gene Details
Entrez Gene ID: | 242285 |
Gene Name: | short chain dehydrogenase/reductase family 16C, member 5 |
Gene Aliases: |
Rdhe2, Scdr9 |
Location: |
Chr.4:3995942-4019663 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sdr16c5 | NM_181989.1 | 2 | 338 | NP_871789.1 |
| AK136508.1 | 1 | 309 | BAE23016.1 | |
| AY277588.1 | 2 | 338 | AAP41074.1 | |
| BC104134.1 | 2 | 257 | AAI04135.1 | |
| BC104135.1 | 2 | 257 | AAI04136.1 |

Genomic Map