Assay Details
Target Gene Details
Entrez Gene ID: | 100129 |
Gene Name: | G protein-coupled receptor 153 |
Gene Aliases: |
1110065N12Rik, AI449320, PGR1 |
Location: |
Chr.4:152273938-152285343 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gpr153 | NM_178406.2 | 2 | 182 | NP_848493.1 |
| AY255610.1 | 1 | 28 | AAO85122.1 | |
| BC029098.1 | 1 | 88 | AAH29098.2 | |
| BC037650.1 | 1 | 181 | AAH37650.1 |
More Information
Set Membership: |
|

Genomic Map