Assay Details
Target Gene Details
Entrez Gene ID: | 100129 |
Gene Name: | G protein-coupled receptor 153 |
Gene Aliases: |
1110065N12Rik, AI449320, PGR1 |
Location: |
Chr.4:152273938-152285343 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gpr153 | NM_178406.2 | 6 | 2115 | NP_848493.1 |
| BC029098.1 | 5 | 2021 | AAH29098.2 | |
| BC037650.1 | 5 | 2114 | AAH37650.1 |
More Information
Set Membership: |
|

Genomic Map