Assay Details
Target Gene Details
Entrez Gene ID: | 81006 |
Gene Name: | G protein-coupled receptor 63 |
Gene Aliases: |
PSP24beta, Psp24-2 |
Location: |
Chr.4:24966541-25009233 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gpr63 | NM_030733.3 | 2 | 614 | NP_109658.2 |
| AK036257.1 | 2 | 615 | BAC29364.1 |
More Information
Set Membership: |
|

Genomic Map