Assay Details
Target Gene Details
Entrez Gene ID: | 73723 |
Gene Name: | SH3 domain binding glutamic acid-rich protein-like 3 |
Gene Aliases: |
1110004L05Rik, AU022188 |
Location: |
Chr.4:134127406-134128753 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sh3bgrl3 | NM_080559.1 | 3 | 507 | NP_542126.1 |
| AK003431.1 | 3 | 488 | BAC25034.1 | |
| BC008110.1 | 3 | 507 | AAH08110.1 |
Target Gene Details
Entrez Gene ID: | 67586 |
Gene Name: | UBX domain protein 11 |
Gene Aliases: |
4930506L07Rik, D4Bwg1540e, Soc, Soci, Ubxd5 |
Location: |
Chr.4:134102512-134127621 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ubxn11 |

Genomic Map