Assay Details
Target Gene Details
Entrez Gene ID: | 21894 |
Gene Name: | talin 1 |
Gene Aliases: |
Tln |
Location: |
Chr.4:43531513-43562583 on Build GRCm38 |
Assay Gene Location: | Within Exon 57 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tln1 | NM_011602.5 | 57 | 8374 | NP_035732.2 |
| AK122420.2 | 57 | 8000 | BAC65702.4 | |
| AK147228.1 | 57 | 7945 | BAE27781.1 | |
| AK170630.1 | 23 | 3417 | BAE41923.1 | |
| AK171322.1 | 23 | 3417 | BAE42391.1 | |
| BC018557.1 | 21 | 3082 | AAH18557.1 | |
| BC121787.1 | 27 | 3886 | ||
| CJ242702.1 | 1 | 260 | ||
| X56123.1 | 57 | 8046 | CAA39588.1 |

Genomic Map