Assay Details
Target Gene Details
Entrez Gene ID: | 70902 |
Gene Name: | lysophosphatidylcholine acyltransferase 2B |
Gene Aliases: |
4921521K07Rik, Aytl1b |
Location: |
Chr.5:107431549-107435039 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lpcat2b | NM_027599.3 | 1 | 362 | NP_081875.1 |
| AK014940.1 | 1 | 363 | BAB29630.1 | |
| AK029548.1 | 1 | 363 | BAC26509.1 | |
| BB572733.1 | ||||
| BC139095.1 | ||||
| BC139096.1 | ||||
| BY714455.1 | 1 | 363 |
More Information
Set Membership: |
|

Genomic Map