Assay Details
Target Gene Details
Entrez Gene ID: | 13120 |
Gene Name: | cytochrome P450, family 4, subfamily b, polypeptide 1 |
Gene Aliases: |
- |
Location: |
Chr.4:115624728-115647705 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyp4b1 | NM_007823.2 | 12 | 1717 | NP_031849.1 |
| AK078882.1 | 10 | 1341 | ||
| AK143890.1 | 12 | 1730 | BAE25585.1 | |
| AK165445.1 | 12 | 1733 | BAE38189.1 | |
| BC008996.1 | 12 | 1721 | AAH08996.1 | |
| D50834.1 | 12 | 1717 | BAA09446.1 |
More Information
Set Membership: |
|

Genomic Map